Learn More
Electron Microscopy Sciences Maxtaform London Finder Grid, 200 mesh Nickel, 127 µm Pitch
SDP
Catalog No. 5026969
Encompass_Preferred
Maxtaform grids with reference patterns are of the highest consistent quality.
Material: Nickel
Pitch: 127 µm
Catalog No. 50-269-69 Supplier Electron Microscopy Sciences Supplier No. LF200NI
Maxtaform grids with reference patterns are of the highest consistent quality.
Material: Nickel
Pitch: 127 µm