Learn More
Electron Microscopy Sciences Maxtaform London Finder Grid, 200 mesh Gold, 127 µm Pitch
SDP
Catalog No. 5026973
Encompass_Preferred
Maxtaform grids with reference patterns are of the highest consistent quality.
Material: Gold
Pitch: 127 µm
Catalog No. 50-269-73 Supplier Electron Microscopy Sciences Supplier No. LF200AU
Maxtaform grids with reference patterns are of the highest consistent quality.
Material: Gold
Pitch: 127 µm