Learn More
Electron Microscopy Sciences Maxtaform London Finder Grid, 400 mesh Copper, 63 µm Pitch
SDP
Catalog No. 5026974
Encompass_Preferred
Maxtaform grids with reference patterns are of the highest consistent quality.
Material: Copper
Pitch: 63 µm
Catalog No. 50-269-74 Supplier Electron Microscopy Sciences Supplier No. LF400CU
Maxtaform grids with reference patterns are of the highest consistent quality.
Material: Copper
Pitch: 63 µm