Learn More
Electron Microscopy Sciences Maxtaform London Finder Grid, 400 mesh Gold, 63 µm Pitch
SDP
Catalog No. 5026970
Encompass_Preferred
Maxtaform grids with reference patterns are of the highest consistent quality.
Material: Gold
Pitch: 63 µm
Catalog No. 50-269-70 Supplier Electron Microscopy Sciences Supplier No. LF400AU
Maxtaform grids with reference patterns are of the highest consistent quality.
Material: Gold
Pitch: 63 µm