Learn More
MSC Proto Hex Bit Socket, 3/8 Drive, 5/16 Hex, 1 ea
Catalog No. 501920288
Encompass
This hex bit socket features a 3/8 in. drive and a 5/16 in. hex size, with an overall length of 6-29/32 in. and a bit length of 5 in. Constructed from chrome alloy steel, the socket has a black oxide finish on the stem and a polished chrome finish on the remainder, offering a long ball end hex bit design suitable for various applications.
  • Hex bit socket type
  • Drive size: 3/8 in.
  • Hex size: 5/16 in.
  • Bit length: 5 in.
  • Overall length: 6-29/32 in.
  • Constructed from chrome alloy steel
  • Finish: Black oxide on stem; polished chrome
  • Design: Long ball end hex bit
  • Not insulated
  • Not non-sparking
  • Not tether capable
Catalog No. 50-192-0288 Supplier MSC Supplier No. 39937784
This hex bit socket features a 3/8 in. drive and a 5/16 in. hex size, with an overall length of 6-29/32 in. and a bit length of 5 in. Constructed from chrome alloy steel, the socket has a black oxide finish on the stem and a polished chrome finish on the remainder, offering a long ball end hex bit design suitable for various applications.
  • Hex bit socket type
  • Drive size: 3/8 in.
  • Hex size: 5/16 in.
  • Bit length: 5 in.
  • Overall length: 6-29/32 in.
  • Constructed from chrome alloy steel
  • Finish: Black oxide on stem; polished chrome
  • Design: Long ball end hex bit
  • Not insulated
  • Not non-sparking
  • Not tether capable