Learn More
MSC Proto Hex Bit Socket, 3/8 Drive, 7/32 Hex, 1 ea
Catalog No. 501920281
Encompass
This hand hex bit socket is engineered for secure and efficient engagement with hex head fasteners. Designed for professional use, it offers a combination of strength and precision for various fastening and loosening tasks.
  • Hex bit socket type
  • Drive size of 3/8 in for compatibility with standard ratchets and drive tools
  • Hex size of 7/32 in for precise fit
  • Bit length of 5 in for extended reach
  • Constructed from durable chrome alloy steel
  • Features a black oxide finish on the stem for corrosion resistance; polished chrome finish
  • Overall length of 6-29/32 in
Catalog No. 50-192-0281 Supplier MSC Supplier No. 39937305
This hand hex bit socket is engineered for secure and efficient engagement with hex head fasteners. Designed for professional use, it offers a combination of strength and precision for various fastening and loosening tasks.
  • Hex bit socket type
  • Drive size of 3/8 in for compatibility with standard ratchets and drive tools
  • Hex size of 7/32 in for precise fit
  • Bit length of 5 in for extended reach
  • Constructed from durable chrome alloy steel
  • Features a black oxide finish on the stem for corrosion resistance; polished chrome finish
  • Overall length of 6-29/32 in