Learn More
Quantum Storage Systems LONG DIVIDER PK6
Supplier:  Quantum Storage Systems DL93030
Encompass_Preferred
Containers can be divided by length and/or width allowing a subdivision down to a 1-1/8 in. square compartment size. Dividers are grey.
Catalog No. 50-752-0861