Learn More
MOVEX INC Long Fume Extractor Arm
Catalog No. NC1850038
Encompass_Preferred
3" Diameter x 60" Long fume extractor arm with polypropylene tubes and 5" diameter polypropylene joints. Ceiling mounted version including a manual damper and an external gas spring.
Catalog No. NC1850038 Supplier MOVEX INC Supplier No. METS150075PP
3" Diameter x 60" Long fume extractor arm with polypropylene tubes and 5" diameter polypropylene joints. Ceiling mounted version including a manual damper and an external gas spring.