Learn More
Slice™ Long-Handled Scraper
Catalog No. 18222855
Change view
Click to view available options
Blade Material:
Zirconium Oxide
Catalog No. Blade Material
18-222-855 Zirconium Oxide
1 options
Catalog No. 18-222-855 Supplier Slice™ Supplier No. 10599
Add to Cart
Add to Cart
The Slice™ 10599 Long-Handled Scraper extends your reach with a sturdy aluminum handle.
  • Installed blade type: Rounded-tip No. 10538
  • Durable metal handle with comfort grip
  • Ambidextrous design
  • Integrated lanyard hole
  • Finger-friendly™ blade scrapes effectively
  • Reduces injuries; lowers costs
  • Chemically inert blades never rusts
  • Oil- and lubricant-free blade
  • 100 percent zirconium oxide blade
  • Blade edge lasts up to 11.2x longer than metal
  • Fewer blade changes = fewer injuries
  • No-tool blade change
Specifications
Product Type Scraper
Blade Material Zirconium Oxide
For Use With (Equipment) Compatible blades: 10538, 10539, 10540
Handle Material Al/Zn Alloy, GFN, PP, TPE, Stainless Steel, Iron
Dimensions (L x W x H) 306.5 x 124.5 x 33.5 mm
Weight (Metric) 286.8 g