Learn More
MSC Vikan Hand Brush, 15.6 in, Stiff, Polyester Bristle, Polypropylene
Catalog No. 502839446
Encompass
This long-handled hand brush features Ultra Safe Technology (UST) with angled filament security units, designed for optimal cleaning in high-risk areas. Its unique bristle system minimizes contamination and bristle loss, making it effective for heavy-duty scrubbing of machines, buckets, containers, and tubs.
  • Features ultra safe technology with angled filament security units.
  • Unique bristle system minimizes contamination and bristle loss.
  • Designed for optimal cleaning in high-risk areas.
  • Suitable for heavy-duty scrubbing.
  • Stiff polyester bristles.
  • Constructed from polypropylene with fiberglass handle.
  • Complies with FDA Regulation CFR 21 for food contact.
  • Max usage temperature for food contact up to 176 °F.
Catalog No. 50-283-9446 Supplier MSC Supplier No. 54807037
This long-handled hand brush features Ultra Safe Technology (UST) with angled filament security units, designed for optimal cleaning in high-risk areas. Its unique bristle system minimizes contamination and bristle loss, making it effective for heavy-duty scrubbing of machines, buckets, containers, and tubs.
  • Features ultra safe technology with angled filament security units.
  • Unique bristle system minimizes contamination and bristle loss.
  • Designed for optimal cleaning in high-risk areas.
  • Suitable for heavy-duty scrubbing.
  • Stiff polyester bristles.
  • Constructed from polypropylene with fiberglass handle.
  • Complies with FDA Regulation CFR 21 for food contact.
  • Max usage temperature for food contact up to 176 °F.