Learn More
Analytical Sales & Services Long Life D2 Lamp & Holder for
Catalog No. 5000804830
Encompass
Long Life D2 Lamp & Holder for Waters M2487 2488 2000 hour warranty
Qty/Pkg: EACH
Catalog No. 50-008-04830 Supplier Analytical Sales & Services Supplier No. 128362
Long Life D2 Lamp & Holder for Waters M2487 2488 2000 hour warranty
Qty/Pkg: EACH