Learn More
Electron Microscopy Sciences EMS Long Molybdenum Boat
SDP
Catalog No. 5026978
Encompass
Thickness: 0.05 mm (0.002") Overall Length is 75 mm. Trough Measures: 50 mm (L) x 5 mm (W) x 1 mm (D)
Catalog No. 50-269-78 Supplier Electron Microscopy Sciences Supplier No. 73818
Thickness: 0.05 mm (0.002") Overall Length is 75 mm. Trough Measures: 50 mm (L) x 5 mm (W) x 1 mm (D)