Learn More
MSC Vikan Mixing Paddle, Polypropylene, 52 inch Length, Blue
Catalog No. 502835843
Encompass
This 52 inch blue polypropylene mixing paddle is a versatile two-in-one tool for mixing and scraping in food processing applications. Its one-piece construction and long handle provide ergonomic access to deep tanks, offering a lightweight alternative to heavier metal paddles and reducing user fatigue.
  • Designed for critical hygiene and food safety environments.
  • Versatile mixing and scraping tool.
  • Ideal for soup, confectionery, dough, and sauce manufacturing.
  • Long handle for ergonomic access to deep tanks and vats.
  • Durable construction for long-lasting daily performance.
  • Easy to clean and maintain for hygiene control.
  • Color-coded for hygienic zoning plans and 5S lean programs.
Catalog No. 50-283-5843 Supplier MSC Supplier No. 72545957
This 52 inch blue polypropylene mixing paddle is a versatile two-in-one tool for mixing and scraping in food processing applications. Its one-piece construction and long handle provide ergonomic access to deep tanks, offering a lightweight alternative to heavier metal paddles and reducing user fatigue.
  • Designed for critical hygiene and food safety environments.
  • Versatile mixing and scraping tool.
  • Ideal for soup, confectionery, dough, and sauce manufacturing.
  • Long handle for ergonomic access to deep tanks and vats.
  • Durable construction for long-lasting daily performance.
  • Easy to clean and maintain for hygiene control.
  • Color-coded for hygienic zoning plans and 5S lean programs.