Learn More
IDEX Health & Science Upchurch LONG PEEK FINGERTIGHT 2PC 10PK
Catalog No. NC1919490
Encompass_Preferred
Idex Fingertight Two-Piece Fitting, Long Thread, Natural PEEK, 1/16 in. OD Tubing, 10-32 Coned, Long Standard Knurled Head Nut; 10 to 32 Coned, For Use With: For 0.06 in. O.D. Tubing, Includes: Ferrule
Catalog No. NC1919490 Supplier IDEX Health & Science Upchurch Supplier No. F330X
Idex Fingertight Two-Piece Fitting, Long Thread, Natural PEEK, 1/16 in. OD Tubing, 10-32 Coned, Long Standard Knurled Head Nut; 10 to 32 Coned, For Use With: For 0.06 in. O.D. Tubing, Includes: Ferrule