Learn More
Genhunter Corporation PerfectWestern, 6-Sectional-Long, Clear
Catalog No. NC9755147
Encompass
6-sectional-long container, clear in color. For longer strips from mini-protein gels. Well Dimensions (LxW): 6 wells each 6-3/16 x 1-9/16 x 1-3/4 inches. Overall Dimensions (LxWxH): 10-1/2 x 6-3/16 x 1-3/4 inches.
Catalog No. NC9755147 Supplier Genhunter Corporation Supplier No. B134
6-sectional-long container, clear in color. For longer strips from mini-protein gels. Well Dimensions (LxW): 6 wells each 6-3/16 x 1-9/16 x 1-3/4 inches. Overall Dimensions (LxWxH): 10-1/2 x 6-3/16 x 1-3/4 inches.