Learn More
Perkin Elmer US LLC Long PFA Secure Nut for Quartz Cyclonic Spray Chambers for NexION 300/350
Catalog No. 502105177
Encompass_Preferred
The Standard (6 mm) PFA Spray Chamber Secure Nut (N8141420) is available separately.
Catalog No. 50-210-5177 Supplier Perkin Elmer US LLC Supplier No. N8145321
The Standard (6 mm) PFA Spray Chamber Secure Nut (N8141420) is available separately.