Learn More
Grainger Parker Pipe Nipple, Brass, 1/2 in MNPT, 1-1/2 in L, 1 ea
Catalog No. 502770269
Encompass_Preferred
This brass pipe nipple is designed for connecting pipe sections in various industrial and commercial plumbing systems. It is constructed from seamless C37700 brass, providing durability for air and non-potable water applications.
  • Manufactured from seamless C37700 brass
  • Rated for operating pressures up to 1,000 psi
  • Operating temperature range of -65°F to 250°F
  • Designed for air and non-potable water systems
  • Threaded on both ends with 1/2 in MNPT connections
  • Includes thread sealant
  • Compliant with SAE J530 and SAE J531 standards
Catalog No. 50-277-0269 Supplier Grainger Supplier No. 13Y790
This brass pipe nipple is designed for connecting pipe sections in various industrial and commercial plumbing systems. It is constructed from seamless C37700 brass, providing durability for air and non-potable water applications.
  • Manufactured from seamless C37700 brass
  • Rated for operating pressures up to 1,000 psi
  • Operating temperature range of -65°F to 250°F
  • Designed for air and non-potable water systems
  • Threaded on both ends with 1/2 in MNPT connections
  • Includes thread sealant
  • Compliant with SAE J530 and SAE J531 standards