Learn More
Grainger Parker Pipe Nipple, Brass, 1/2 in, 2 1/2 in, 1,000 psi
Catalog No. 502770678
Encompass_Preferred
This precision-machined brass pipe nipple is designed for reliable, leak-free connections between pipes, fittings, and instruments in various systems. It features a seamless construction and is threaded on both ends to facilitate secure installations using appropriate thread sealants.
  • 1/2 in. nominal pipe size
  • 2-1/2 in. overall length
  • Constructed from C37700 brass for durability and corrosion resistance
  • Rated for a maximum operating pressure of 1,000 psi
  • Operating temperature range of -65°F to 250°F
  • Suitable for use with air and non-potable water
  • Complies with SAE J530 and SAE J531 standards
Catalog No. 50-277-0678 Supplier Grainger Supplier No. 13Y792
This precision-machined brass pipe nipple is designed for reliable, leak-free connections between pipes, fittings, and instruments in various systems. It features a seamless construction and is threaded on both ends to facilitate secure installations using appropriate thread sealants.
  • 1/2 in. nominal pipe size
  • 2-1/2 in. overall length
  • Constructed from C37700 brass for durability and corrosion resistance
  • Rated for a maximum operating pressure of 1,000 psi
  • Operating temperature range of -65°F to 250°F
  • Suitable for use with air and non-potable water
  • Complies with SAE J530 and SAE J531 standards