Learn More
Electron Microscopy Sciences Earth-Friendly Long Handle Sampling Spoon, 1.25 ml
SDP
Catalog No. 501927067
Encompass_Preferred
Long-handled spoons are ideal for acquiring samples from bottles, jars and other confined areas and helping to avoid personal contact with sample materials. A flat edge on the rim of each bowl allows fast leveling to collect and measure small amounts with accuracy. Spoon is 17.8cm (7") overall length. Non-sterile and translucent in color.
Capacity: 1.25 ml
Catalog No. 50-192-7067 Supplier Electron Microscopy Sciences Supplier No. 72134
Long-handled spoons are ideal for acquiring samples from bottles, jars and other confined areas and helping to avoid personal contact with sample materials. A flat edge on the rim of each bowl allows fast leveling to collect and measure small amounts with accuracy. Spoon is 17.8cm (7") overall length. Non-sterile and translucent in color.
Capacity: 1.25 ml