Learn More
New Era Enterprises LONG TIP NMR PIPETS 100/PK
Catalog No. NC9354988
Encompass_Preferred
Pipet, Long Tip, for 3mm x 7 in. Tube7.5 in.tip 10.5 in. OAL
Catalog No. NC9354988 Supplier New Era Enterprises Supplier No. NE201B
Pipet, Long Tip, for 3mm x 7 in. Tube7.5 in.tip 10.5 in. OAL