Learn More
Wilmad Labglass Long-Tip Pipet,13-1/4"OAL,Bx/100 for 7",8",9" Tubes
Catalog No. NC1087265
Encompass
Use the Long-Tip Sample Transfer Pipets for easy addition or removal of liquid samples from the longest NMR and EPR sample tubes, volumetric flasks with long, narrow necks and chromatography columns.
Catalog No. NC1087265 Supplier Wilmad Labglass Supplier No. 803A
Use the Long-Tip Sample Transfer Pipets for easy addition or removal of liquid samples from the longest NMR and EPR sample tubes, volumetric flasks with long, narrow necks and chromatography columns.