Learn More
New England Biolabs, Inc. LongAmp Hot Start Taq DNA Polymerase – 500 units
SDP
Catalog No. 50591075
Encompass
LongAmp Hot Start Taq DNA Polymerase is a unique blend of aptamer-based Hot Start Taq and Deep Vent DNA Polymerases. The aptamer-based inhibitor binds reversibly to the enzyme, inhibiting polymerase activity at temperatures below 45C, but releases the enzyme during normal PCR cycling conditions. This permits assembly of PCR reactions at room temperature. An advantage of the aptamer-based hot start mechanism is that it does not require a separate high temperature incubation step to activate the enzyme. The 3' to 5' exonuclease activity of Deep Vent DNA Polymerase increases the fidelity and robust amplification of Hot Start Taq DNA Polymerase. LongAmp Hot Start Taq DNA Polymerase offers two-fold higher fidelity than Hot Start Taq DNA Polymerase alone. A wide range of PCR products can be generated; up to 30 kb from lambda or human genomic DNA.
Catalog No. 50-591-075 Supplier New England Biolabs, Inc. Supplier No. M0534S
LongAmp Hot Start Taq DNA Polymerase is a unique blend of aptamer-based Hot Start Taq and Deep Vent DNA Polymerases. The aptamer-based inhibitor binds reversibly to the enzyme, inhibiting polymerase activity at temperatures below 45C, but releases the enzyme during normal PCR cycling conditions. This permits assembly of PCR reactions at room temperature. An advantage of the aptamer-based hot start mechanism is that it does not require a separate high temperature incubation step to activate the enzyme. The 3' to 5' exonuclease activity of Deep Vent DNA Polymerase increases the fidelity and robust amplification of Hot Start Taq DNA Polymerase. LongAmp Hot Start Taq DNA Polymerase offers two-fold higher fidelity than Hot Start Taq DNA Polymerase alone. A wide range of PCR products can be generated; up to 30 kb from lambda or human genomic DNA.