Learn More
New England Biolabs, Inc. LongAmp Taq DNA Polymerase – 500 units
SDP
Catalog No. 50994936
Encompass
LongAmp Taq DNA Polymerase is a unique blend of Taq and Deep Vent DNA Polymerases. The 3' to 5' exonuclease activity of Deep Vent DNA Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. LongAmp Taq DNA Polymerase offers two fold higher fidelity than Taq DNA Polymerase alone. A wide range of PCR products can be generated, up to 30 kb from lambda DNA or from human genomic DNA.
Catalog No. 50-994-936 Supplier New England Biolabs, Inc. Supplier No. M0323S
LongAmp Taq DNA Polymerase is a unique blend of Taq and Deep Vent DNA Polymerases. The 3' to 5' exonuclease activity of Deep Vent DNA Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. LongAmp Taq DNA Polymerase offers two fold higher fidelity than Taq DNA Polymerase alone. A wide range of PCR products can be generated, up to 30 kb from lambda DNA or from human genomic DNA.