Learn More
New England Biolabs, Inc. LongAmp Taq PCR Kit – 100 reactions (50 µl vol)
SDP
Catalog No. 50995156
Encompass
The LongAmp Taq PCR Kit features LongAmp Taq DNA Polymerase, which is a unique blend of Taq and Deep Vent DNA Polymerases. The 35 exonuclease activity of Deep Vent Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. LongAmp Taq DNA Polymerase offers two fold higher fidelity than Taq Polymerase alone. The LongAmp Taq PCR Kit is supplied with LongAmp Taq Reaction Buffer, which is optimized to give highest PCR yields up to 30 kb. The kit can generate a wide range of PCR products, up to 30 kb from plasmid or from human genomic DNA.
Catalog No. 50-995-156 Supplier New England Biolabs, Inc. Supplier No. E5200S
The LongAmp Taq PCR Kit features LongAmp Taq DNA Polymerase, which is a unique blend of Taq and Deep Vent DNA Polymerases. The 35 exonuclease activity of Deep Vent Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. LongAmp Taq DNA Polymerase offers two fold higher fidelity than Taq Polymerase alone. The LongAmp Taq PCR Kit is supplied with LongAmp Taq Reaction Buffer, which is optimized to give highest PCR yields up to 30 kb. The kit can generate a wide range of PCR products, up to 30 kb from plasmid or from human genomic DNA.