Learn More
Cayman Chemical Long-chain Cis/Trans Isomer Fatty Acid Methyl Ester Mixture, 5 mL
SDP
Catalog No. 502989082
Encompass_Preferred
A mixture of long-chain cis/trans FAMEs; intended for use as a reference standard for the detection of these long-chain cis/trans isomer fatty acids in various sample types by GC-FID or GC-MS
Catalog No. 50-298-9082 Supplier Cayman Chemical Supplier No. 293635 ML
A mixture of long-chain cis/trans FAMEs; intended for use as a reference standard for the detection of these long-chain cis/trans isomer fatty acids in various sample types by GC-FID or GC-MS