Learn More
LONGER PUMP Pumps/ peristaltic pumps/ Longer Pump OEM pump T60-S2&WX10-14, Multi-control Modes, Flow Rate: ≤24ml/min, Suitable for OEM Equipment, Soleplate or Panel Installation
Supplier:  LONGER PUMP 49/67
Encompass_Preferred
Pumps/ peristaltic pumps/ Longer Pump OEM pump T60-S2&WX10-14SpecificationSpeed: less than 60rpmSpeed resolution: 0.1rpmMax flowrate 24ml/mintubing: ID less than 3.17mm, wall thickness 0.8mm-1.0mmexternal control module: 0-5VPower supply: DC11.4V-15.2V/10V
Catalog No. NC2956829