Learn More
LONGER PUMP Pumps/ Syringe pumps/ Longer Pump dLSP 501 uC +1*dLSP 501W,Split-type Syringe Pump, Flow Rate: 0.0048nL/min-95.82mL/min, Up to 32 Channels
Supplier:  LONGER PUMP 31/67
Encompass_Preferred
Pumps/ Syringe pumps/ Longer Pump dLSP 501 uC +1*dLSP 501WSpecification- Work mode: - Infusion only - Withdrawal only - Infusion/withdrawal - Withdrawal/infusion - Programming- Stroke of the drive unit: 95mm- Pusher advance per microstep: 0.098μm/ustep- Linear speed: 0.625μm/min-143.75mm/min- Linear speed resolution: 0.625μm/min- Linear travel accuracy: ≤ ±0.35%- Linear travel CV: 0.03%- Linear force (Max.): 100N- Syringe: 5μL-60mL- Programming function: - Parameter configured based on workflow on PC. - PC method programmer: infusion, withdrawal, delay, repeat. - PC control software: constant flow rate, ramp up/down, delay, repeat- Controller dimensions (L × W × H): 187mm × 123mm × 39mm- Controller weight: 0.6kg- Power supply for controller: AC 90V-260V/30W- Drive unit dimensions (L × W × H): 235mm × 103mm × 91mm- Drive unit weight: 1.65kg- Power supply for drive unit: DC 24V/5W
Catalog No. NC2956925