Learn More
LONGER PUMP Pumps/ peristaltic pumps/ Longer Pump dPOFLEX GP01 with pump head GPH02, Industrial Peristaltic Pump, Stainless Steel Housing, Flow Rate: 0.001-17L/min, IP66, meets 21CFR Part 11 GMP requirements
Supplier:  LONGER PUMP 27/67
Encompass_Preferred
Pumps/ peristaltic pumps/ Longer Pump dPOFLEX GP01 with pump head GPH02,Speicification1. Model: dPOFLEX GP01, BP012. Speed: 0.1-265rpm3. Resolution: 0.1rpm4. Modes: Flow rate, dispensing, calibration5. Flow: Continuous, displays rate/speed6. Rate: 1.1ml/min-17.0L/min7. Dispensing: Set volume, cycles, interval8. Single: 1.0mL-274.9L9. Cycles: 1-999910. Interval: 55-9999s11. Control: Keypad, foot switch, external signal, communication12. Comm: RS485, Ethernet, Modbus RTU, TCP/IP, Profibus DP13. External: Speed (0-10V, 4-20mA), start/stop, direction (5V-24V)14. Output: Pump Speed (0-10V, 4-20mA)15. Signature: FDA 21CFR Part 1116. Verification: IQ/OQ17. Cert: CE, UKCA, EN 61010-1, EN IEC 61010-2, EMC EN IEC 61000-618. Dim: 260x407x298mm (GP01), 260x407x290mm (BP01)19. Power: AC100-240V 50/60Hz20. Condition: 5°C-40°C, humidity ≤100%21. IP: IP6622. Weight: ≤17.6kg (GP01), ≤16.6kg (BP01)
Catalog No. NC2956857
Add to Cart
Request a Quote