Learn More
LONGER PUMP Pumps/ peristaltic pumps/ Longer Pump dPOFLEX GP02 with pump head KZ35-13-D, Flow Rate: 1ml/min-9L/min, IP66, supporting fieldbus communication control, digital signal control, analog signal control
Supplier:  LONGER PUMP 28/67
Encompass_Preferred
Pumps/ peristaltic pumps/ Longer Pump dPOFLEX GP02Specification- Housing Material: Cast aluminum- Speed Range: 1rpm-300rpm, CW/CCW- Speed Resolution: 0.1rpm- Work Mode: Flow rate, dispensing, calibration- Flow Rate Setting Range: 1.0mL/min-9.0L/min- Volume of Single Dispense: 1.0mL-225L- Dispensing Cycles: 1-9999 and unlimited- Interval Time: 1s-9999s- Control Mode: Keypad, external signal, communication, foot switch- External Control: 0-5V, 0-10V, 0-10kHz, 4-20mA; Start/stop, direction: 5V-24V logic level or relay- Status Output: Running/stop, fault: relay signal- Power Status: Stops or runs in flow rate or dispensing mode- Electronic Signature: FDA 21CFR Part 11- Certificate: CE, UKCA- Power Supply: AC100-240V/115W 50/60Hz- Weight: ≤10.4kg
Catalog No. NC2956860
Add to Cart
Request a Quote