Learn More
LONGER PUMP Pumps/ peristaltic pumps/ Longer Pump G100-1L&BPH01, Die-cast Aluminum Housing, Flow Rate: 0.15µL/min-3250mL/min, IP66, continuous transfer, timed transfer, volume transfer, CE Certified
Catalog No. NC2956908
Encompass_Preferred
Pumps/ peristaltic pumps/ Longer Pump G100-1L&BPH01Specification- Speed range: 0, 0.01-150 (rpm)- Speed resolution: 0.01rpm (< 10rpm), 0.1rpm (10rpm ≤ speed < 100rpm)- Flow rate range: 0.15uL/min-750mL/min- Work mode: Continuous transfer, timed transfer, volume transfer- External control: Start/stop control, direction control: logic level signal or switch signal, momentary or maintained trigger mode can be configured. Speed control: 0-5V/0-10V/4-20mA/0-10kHz with uniform interface, speed range/signal range can be configured- Communication control: RS485 interface, Modbus RTU and Longer Pump OEM protocol- Prime: Fast filling or emptying at full speed- Certificate: CE, UKCA- Dimension (LxWxH): 240*142*152 (mm)- Power supply: AC100-240V 50Hz/60Hz- Power consumption: 25W- Working condition: Environment temperature: 0°C-40°C, relative humidity: 10%-90%- IP rating: IP66- Weight: 3.64kg
Catalog No. NC2956908 Supplier LONGER PUMP Supplier No. 29/67
Pumps/ peristaltic pumps/ Longer Pump G100-1L&BPH01Specification- Speed range: 0, 0.01-150 (rpm)- Speed resolution: 0.01rpm (< 10rpm), 0.1rpm (10rpm ≤ speed < 100rpm)- Flow rate range: 0.15uL/min-750mL/min- Work mode: Continuous transfer, timed transfer, volume transfer- External control: Start/stop control, direction control: logic level signal or switch signal, momentary or maintained trigger mode can be configured. Speed control: 0-5V/0-10V/4-20mA/0-10kHz with uniform interface, speed range/signal range can be configured- Communication control: RS485 interface, Modbus RTU and Longer Pump OEM protocol- Prime: Fast filling or emptying at full speed- Certificate: CE, UKCA- Dimension (LxWxH): 240*142*152 (mm)- Power supply: AC100-240V 50Hz/60Hz- Power consumption: 25W- Working condition: Environment temperature: 0°C-40°C, relative humidity: 10%-90%- IP rating: IP66- Weight: 3.64kg