Learn More
LONGER PUMP Pumps/ peristaltic pumps/ Longer Pump Pump head KZ25, Stainless Steel Rotor, Flow Rate: ≤6000ml/min, Easy Tubing Change, Self-lubrication Function
Supplier:  LONGER PUMP 7/67
Encompass_Preferred
Pumps/ peristaltic pumps/ Longer Pump Pump head KZ25Applications:Laboratory, analytical instrument, IndustryProduct Instruction:The pump head consists of stainless steel rotor, transparent PC housing and PPS compression block. The pump running state is visible through the transparent housing. The PPS compression block has self-lubrication function to reduce the tubing wear. And the tubing could be loaded and removed easily.
Catalog No. NC2956843