Learn More
LONGER PUMP Pumps/ peristaltic pumps/ Longer Pump JL350-2J, Large Flow Rates, AC Motor, Flow Rate: 1.3L/min-35L/min, Variable Frequency Control, IP55, Ideal for Industrial and High-capacity Applications
Supplier:  LONGER PUMP 18/67
Encompass_Preferred
Pumps/ peristaltic pumps/ Longer Pump JL350-2JSpecification- Speed: 30rpm-350rpm, CW/CCW- Speed resolution: 0.6rpm- Control mode: Keypad control or external signal control- Display: LCD for running information- Speed control: 0-5V, 0-10V, 4-20mA linear control- Start/stop signal input: Switch signal control- Direction signal input: Switch signal control- Communication function: RS485 interface, Modbus RTU protocol- Memory function: Storing the running parameter automatically- Dimension (L × W × H): 417 × 401 × 321 (mm)- Power supply: AC 220V ±10% / 400W- Operating temperature: 0 to 40°C- Relative humidity: <80%- IP rating: IP55 (excluding frequency converter)- Weight: 32kg
Catalog No. NC2956918
Add to Cart
Request a Quote