Learn More
LONGER PUMP Pumps/ peristaltic pumps/ Longer Pump L100-1F with pump head YZ1515X-A, Touch Screen, Flow Rate: 0.15µl/min-500ml/min, Programmable, Multiple Control Modes, IP31, Lab and Industrial Applications
Catalog No. NC2956868
Encompass_Preferred
Pumps/ peristaltic pumps/ Longer Pump L100-1F&YZ1515X-ASpecifications- Speed: <100rpm- Speed resolution: 0.1rpm- Flow rate: 0.15uL/min-500mL/min- Dispensing volume: 0.1mL-9999L, accuracy of ±2%- Work mode: Programming- Programming function: Running control module: constant, ramp up/down, stepped up/down, sine, constant dispensing, incremental/decremental dispensing. Logic control module: direction, delay, event trigger, status output, pause, jump, loop, stop. Configure multi-step program with multiple modules for complex application- External control: 0-5V, 0-10V, 4-20mA or 0-10kHz - Dispensing volume setting range: 0.001uL-9999L- Dispensing time and interval time: 0.5s-9999s- Dispensing cycle: 1-999999 (0 for unlimited)- Delay time setting range: 0.5s-9000s- Power Supply: AC100V-240V, 50/60Hz- Weight: 5.0kg
Catalog No. NC2956868 Supplier LONGER PUMP Supplier No. 12/67
Pumps/ peristaltic pumps/ Longer Pump L100-1F&YZ1515X-ASpecifications- Speed: <100rpm- Speed resolution: 0.1rpm- Flow rate: 0.15uL/min-500mL/min- Dispensing volume: 0.1mL-9999L, accuracy of ±2%- Work mode: Programming- Programming function: Running control module: constant, ramp up/down, stepped up/down, sine, constant dispensing, incremental/decremental dispensing. Logic control module: direction, delay, event trigger, status output, pause, jump, loop, stop. Configure multi-step program with multiple modules for complex application- External control: 0-5V, 0-10V, 4-20mA or 0-10kHz - Dispensing volume setting range: 0.001uL-9999L- Dispensing time and interval time: 0.5s-9999s- Dispensing cycle: 1-999999 (0 for unlimited)- Delay time setting range: 0.5s-9000s- Power Supply: AC100V-240V, 50/60Hz- Weight: 5.0kg