Learn More
LONGER PUMP Pumps/ peristaltic pumps/ Longer Pump L100-1S-2 with pump head YZ1515X-A, precise speed resolution 0.01rpm, Flow Rate: 0.00015-500ml/min, CE Certified, Ideal for Laboratory and Light Industry
Supplier:  LONGER PUMP 20/67
Encompass_Preferred
Pumps/ peristaltic pumps/ Longer Pump 100-1S-2&YZ1515X-ASpecifications- Speed: 0.01rpm-100rpm- Speed resolution: 0.01rpm - Control mode: Keypad control, external signal control and communication control- Work mode: Speed control mode and flow rate mode- Flow rate setting range: 0.01µL/min-999L/min- External control: Start/stop control, direction control, speed control - Communication control: RS485 interface, Longer protocol or Modbus protocol- Timing function: Set the running time- Dimension (L × W × H): 232 × 142 × 149 (mm)- Power supply: AC100V-240V/25W, 50/60Hz- Operating temperature: 0 to 40°C- Relative humidity: <80%- IP rating: IP31- Weight: 2.38kg
Catalog No. NC2956881