Learn More
LONGER PUMP Pumps/ Syringe pumps/ Longer Pump LSP01-3A Single Channel Syringe Pump, High Accuracy, Flow Rate: 0.001μl/min-43.349ml/min, Suitable for Laboratory Applications
Supplier:  LONGER PUMP 34/67
Encompass_Preferred
Pumps/ Syringe pumps/ Longer Pump LSP01-3ASpecifications- Work mode: Infusion- Stroke of pump: 140mm- Pusher advance per microstep: 0.156μm- Linear speed: 5μm/min-65mm/min- Linear speed resolution: 5μm/min- Linear travel accuracy: ≤ ±0.5% when travel >30% of pump stroke- Linear force (max.): >90N- Running parameters setting: Infusion volume, infusion time, etc.- Display setting: Display volume, flow rate or linear speed- Power-off memory: Storing the running parameters automatically- Status signal output: 2 output signals (OC gate signal) to indicate start/stop and direction- Control signal input: Falling edge or TTL signal to control Start/stop- Communication interface: RS485- Dimensions (L × W × H): 280 × 210 × 140 mm- Weight: 3.6kg- Power supply: AC 196V-240V/20W- Operating temperature: 0 to 40°C- Relative humidity: <80%
Catalog No. NC2956920