Learn More
LONGER PUMP Pumps/ peristaltic pumps/ Longer Pump/ Piston pump WT3000-1FB&MS204 Flow Rate: 85.7-2571.4ml/min, Up to 1 Channel, Max Pressure: 1.4MPa, Suitable for Lab and Industry Applications
Supplier:  LONGER PUMP 37/67
Encompass_Preferred
Pumps/ peristaltic pumps/ Longer Pump WT3000-1FB&MS204Specification- Precision: - Dispensing 100% of total volume: - CV<0.1% (Pump Volume<500μL) - CV<0.05% (Pump Volume ≥ 500μL) - Dispensing 10% of total volume: - CV<0.5% (Pump Volume<500μL) - CV<0.1% (Pump Volume ≥ 500μL) - Dispensing 1% of total volume: - CV<2% (Pump Volume<500μL) - CV<1% (Pump Volume ≥ 500μL)- Accuracy: ≥99.5% at full dispensing- Expected life: - 10 Million Cycles with Distilled Water - 5 Million Cycles with Crystallizable solution- System pressure: ≤0.68Mpa (Depending on model)- Full stroke: 12.7mm- Resolution: - 2000 steps: 0.00635mm/step - 4000 steps: 0.003175mm/step- Pump speed: - Pump volume ≤1mL: the min. time for full stroke is 0.8s - Pump volume of 2.5mL: the min. time for full stroke is 1s - Pump volume of 5mL: the min. time for full stroke is 1.2s- Dimension (L × W × H): 61 × 45 × 132 (mm)- Weight: 0.51kg
Catalog No. NC2956919