Learn More
LONGER PUMP Pumps/ peristaltic pumps/ Longer Pump WT600-1F&KZ25-13, Flow Rate: 0.7-6000ml/min, Up to 3 Channels, CE Certified, Ideal for Laboratory and Light Industry
Catalog No. NC2956916
Encompass_Preferred
Pumps/ peristaltic pumps/ Longer Pump WT600-1F&KZ25-13Specification- Speed: 10rpm-600rpm- Speed resolution: 1rpm- Control mode: Control panel, external signal control and communication control are available- Work mode: Flowrate mode and dispensing mode- Dispensing volume: 0.1mL-99.9L- Copy number: 1 to 9999, "0" means unlimited cycle- Pause time: 0.1s to 99.9min, resolution is 0.1s- Back suction circle: 0 to 9.9 circle, resolution is 0.1 circle- External control: Start/stop, direction and speed control (4-20mA, 0-5V, 0-10V, 0-10kHz external control module optional)- Speed output: 166.7Hz-10kHz corresponding to 10-600rpm- Communication interface: RS485- Power-off memory: Return to previous status when powered on- Dimension (L×W×H): 285 x 207 x 180 (mm)- Power supply: AC 110V±20%/140W- Operating temperature: 0 to 40°C- Relative humidity: <80%- IP rating: IP31- Weight: 5.2kg
Catalog No. NC2956916 Supplier LONGER PUMP Supplier No. 25/67
Pumps/ peristaltic pumps/ Longer Pump WT600-1F&KZ25-13Specification- Speed: 10rpm-600rpm- Speed resolution: 1rpm- Control mode: Control panel, external signal control and communication control are available- Work mode: Flowrate mode and dispensing mode- Dispensing volume: 0.1mL-99.9L- Copy number: 1 to 9999, "0" means unlimited cycle- Pause time: 0.1s to 99.9min, resolution is 0.1s- Back suction circle: 0 to 9.9 circle, resolution is 0.1 circle- External control: Start/stop, direction and speed control (4-20mA, 0-5V, 0-10V, 0-10kHz external control module optional)- Speed output: 166.7Hz-10kHz corresponding to 10-600rpm- Communication interface: RS485- Power-off memory: Return to previous status when powered on- Dimension (L×W×H): 285 x 207 x 180 (mm)- Power supply: AC 110V±20%/140W- Operating temperature: 0 to 40°C- Relative humidity: <80%- IP rating: IP31- Weight: 5.2kg