Learn More
Cayman Chemical Epimedin B, Purity: ≥98%, CAS: 110623-73-9, 50 mg
SDP
Catalog No. 502989115
Encompass_Preferred
A flavonoid glycoside; scavenges DPPH radicals in a cell-free assay (IC50 = 107.9 µM); increases proliferation of UMR-106 osteoblasts by 24.9 and 36.3% at 10 and 100 µM, respectively; prevents prednisolone-induced osteoporosis in zebrafish at 1 µM; inhibits TPA-induced ear edema in mice (ED50 = 114 nmol/ear)
Catalog No. 50-298-9115 Supplier Cayman Chemical Supplier No. 2969450 MG
A flavonoid glycoside; scavenges DPPH radicals in a cell-free assay (IC50 = 107.9 µM); increases proliferation of UMR-106 osteoblasts by 24.9 and 36.3% at 10 and 100 µM, respectively; prevents prednisolone-induced osteoporosis in zebrafish at 1 µM; inhibits TPA-induced ear edema in mice (ED50 = 114 nmol/ear)