Learn More
Abcam Lonidamine, He x okinase inhibitor, 5MG
Catalog No. 502706234
Encompass_Preferred
MW 321.2 Da, Purity >98%. Hexokinase inhibitor (IC₅₀ = 850 μM). Anticancer, antiparasitic and antispermatogenic activity in vivo. Blocks open CFTR Cl- channels in vitro. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6234 Supplier Abcam Supplier No. AB1424425MG
MW 321.2 Da, Purity >98%. Hexokinase inhibitor (IC₅₀ = 850 μM). Anticancer, antiparasitic and antispermatogenic activity in vivo. Blocks open CFTR Cl- channels in vitro. Orally active.
The product is subject to the following: Abcam Restricted Use Statement