Learn More
Aviva Systems Biology Corp LONRF1 Antibody - N-terminal region (ARP43416_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43416_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against LONRF1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human LONRF1. Alternate names: FLJ23749, RNF191ApplicationIHC, WB. Species reactivity: Human, Mouse, Rat
Catalog No. 50-014-58524