Learn More
Aviva Systems Biology Corp LONRF2 Antibody - middle region (ARP43498_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43498_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LONRF2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LONRF2. Alternate names: FLJ45273, MGC126711, MGC126713, RNF192ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58552