Learn More
Cayman Chemical Lonurus artmsa Lonurn 100mg
SDP
Catalog No. 502932784
Encompass_Preferred
An alkaloid that has diverse biological activities; reduces infarct size and collagen deposition and inhibits apoptosis of cardiomyocytes in a coronary artery ligation-induced rat model of myocardial infarction at 15 mg/kg per day; increases hepatic levels of SOD and GSH, reduces serum ALT and AST levels, and inhibits hepatic fibrosis in a diet-induced mouse model of NASH at 200 mg/kg per day; inhibits LPS-induced increases in TNF-α, IL-1β, and IL-6 levels in the serum of broiler chicks at 120 mg/kg
Catalog No. 50-293-2784 Supplier Cayman Chemical Supplier No. 26850100 MG
An alkaloid that has diverse biological activities; reduces infarct size and collagen deposition and inhibits apoptosis of cardiomyocytes in a coronary artery ligation-induced rat model of myocardial infarction at 15 mg/kg per day; increases hepatic levels of SOD and GSH, reduces serum ALT and AST levels, and inhibits hepatic fibrosis in a diet-induced mouse model of NASH at 200 mg/kg per day; inhibits LPS-induced increases in TNF-α, IL-1β, and IL-6 levels in the serum of broiler chicks at 120 mg/kg