Learn More
Description
- Gold standard
- Formulated for use with PAGEr™ precast gels
- Clear or colorless aqueous solution
- Miscible in water
- Prepared with high quality reagents and use 18megohm water
- Filtered using 0.2μ filter
- Ready-to-use
Specifications
Specifications
| CAS | 56-40-6, 7732-18-5 |
| Concentration | 10X |
| For Use With (Application) | Protein Electrophoresis |
| Physical Form | Liquid |
| Quantity | 1 L |
| Solution Type | Buffer |
| Formulation | Tris Glycine Buffer, 10X with 0.25M Tris base, 1.92M Glycine |
| Grade | Electrophoresis |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.