Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AccuGENE™ 10X Tris-Glycine-SDS Buffers, Lonza™

Catalog No. BMA50880
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BMA50880 1 L
BMA50882 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BMA50880 Supplier Lonza Inc Supplier No. 50880
Add to Cart
Add to Cart

For use with denaturing protein electrophoresis

  • Gold standard
  • Formulated for use with PAGEr™ precast gels
  • Clear or colorless aqueous solution
  • Miscible in water
  • Prepared with high quality reagents and use 18megohm water
  • Filtered using 0.2μ filter
  • Ready-to-use

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Protein Sample Loading Buffer
Concentration 10X
Physical Form Liquid
Quantity 1 L
Formulation Tris Glycine-SDS Buffer, 10X with 0.25M Tris base, 1.92M Glycine, 1% SDS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.