Learn More
Description
AccuGENE™ Molecular Biology Buffers are ready-to-use solutions ideal for a wide range of molecular biology applications. Each reagent is prepared with 18 megOhm water, filtered using a 0.2 micron filter, and filled into sterile bottles.
Specifications
Specifications
| Chemical Name or Material | 1X TE Buffer |
| CAS | 6381-92-6, 77-86-1 |
| Concentration | 1X |
| For Use With (Application) | Molecular biology applications |
| pH | 7.4 |
| Physical Form | Aqueous Solution |
| Quantity | 500 mL |
| Solution Type | Buffer |
| Formulation | Tris, 0.001M EDTA (Disodium salt) 0.01M |
| Grade | Molecular Biology |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.