Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ AccuGENE™ Media

Catalog No. BMA51221 Shop All Lonza, Inc Products
Change view
Click to view available options
Accugene, Super Broth; 1L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
BMA51221 Accugene, Super Broth; 1L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BMA51221 Supplier Lonza™ Supplier No. 51221

missing translation for 'validMainframeMsg'

  • Ready-to-use solutions save prep time
  • Each reagent prepared with 18 megOhm water, filtered using a 0.2μm filter, and filled into sterile bottles
  • Manufactured according to strict quality control standards to ensure lot-to-lot consistency
  • No detectable DNase, RNase, or protease activities unless otherwise noted
  • Storage Conditions: 18°C–24°C

TRUSTED_SUSTAINABILITY

Specifications

Media Type Terrific Broth
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.