Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ AccuGENE™ PBS

Catalog No. BMA51226 Shop All Lonza, Inc Products
Change view
Click to view available options
Concentration:
10X
1X
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
BMA51226 10X
BMA51225 1X
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BMA51226 Supplier Lonza™ Supplier No. 51226
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ready to use solutions ideal for a wide range of molecular biology applications

AccuGENE Molecular Biology Buffers are ready-to-use solutions ideal for a wide range of molecular biology applications. Each reagent is prepared with 18 megOhm water, filtered using a 0.2 micron filter, and filled into sterile bottles.

  • Reliable - Manufactured according to strict quality control standards to ensure lot-to-lot consistency.
  • High Quality - No detectable DNase, RNase, or protease activities unless otherwise noted.
  • Efficient - Ready-made solutions eliminate experiment preparation time.
  • Flexible - Customized solutions are available to meet individual needs

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10X
For Use With (Application) Ideal for a wide range of molecular biology applications
pH 7.4
Quantity 1000mL
Solution Type Buffer
Formulation KH2PO4 0.017M, Na2HPO4 0.05M, NaCl 1.5M
Grade Not Available
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.