Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AccuGENE™ TAE Buffers, Lonza™

Catalog No. BMA51216 Shop All Lonza, Inc Products
Change view
Click to view available options
Quantity:
1 L
4 L
Concentration:
10X
50X
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
BMA51216 1 L 50X
BMA50844 1 L 10X
BMA50841 4 L 10X
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BMA51216 Supplier Lonza Inc Supplier No. 51216
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Formulated to match Lonza's Reliant™ and Latitude™ gels

  • Ready-to-use
  • Prepared with high quality reagents and use 18megohm water
  • Filtered using 0.2μ filter
  • Available in 10X and 50X strength
  • Manufactured according to strict quality control standards to ensure consistency
  • Customized solutions are available to meet individual needs

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DNA Sample Loading Buffer-TAE
CAS 64-19-7
Concentration 50X
For Use With (Application) Agarose Gel Electrophoresis, Polyacrylamide Gel Electrophoresis
Physical Form Liquid
Quantity 1 L
Solution Type Buffer
Formulation TAE Buffer, 50X with 2M Tris acetate, 0.05M EDTA
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.