Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ BioWhittaker™ Alpha MEM Eagle Medium

Catalog No. BW12169F Shop All Lonza, Inc Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BW12-169F 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BW12-169F Supplier Lonza™ Supplier No. 12169F

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used to culture bone marrow cells and amniotic cells for chromosome analysis. BioWhittaker™ Alpha MEM Eagle Medium are manufactured in accordance with cGMP regulations.

Differs from standard MEM by containing the following unique components:

  • Vitamin B12
  • Ascorbic acid
  • Non-essential amino acids
  • Sodium pyruvate
  • Lipoic acid
  • D-Biotin
TRUSTED_SUSTAINABILITY

Specifications

Sterility Sterile
Sterilization Method Sterile-filtered through two consecutive 0.2 μm filters
Without Additives No Glutamine, No Deoxyribonucleosides, No Ribonucleosides
Quantity 500 mL
pH 6.91 to 7.27
Content And Storage 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.