Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ BioWhittaker™ Cell Culture Reagents

Catalog No. BW17613E Shop All Lonza, Inc Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BW17-613E 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BW17-613E Supplier Lonza™ Supplier No. 17613E

missing translation for 'validMainframeMsg'

Manufactured in accordance with cGMP regulations and cell culture-tested to assure compatibility

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15° to 30°C
Product Type Sodium Bicarbonate
Sterility Sterile
pH 7.5 to 8.02
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.